Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UCH37 Monoclonal Antibody (854CT5.5.6)
GREENER_CHOICE

Catalog No. PIMA518114
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518114 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518114 Supplier Invitrogen™ Supplier No. MA518114
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UCH37 Monoclonal Antibody for Western Blot

UCH-L5 (ubiquitin carboxyl-terminal hydrolase isozyme L5), also known as UCH37, is a 329 amino acid protein that functions to edit polyubiquinated protein substrates. Since UCH-L5 has the potential to rescue ubiquinated proteins, including oncogenic proteins, from proteasomal degradation, it is likely that deregulation of UCH-L5 may affect tumor growth. Through associations with Smad7, UHC-L5 can dramatically upregulate TGFβ-dependent gene expression by de-ubiquinating and stabilizing TGFβ RI. Also, since overexpression of UCH-L5 and other deubiquitinating enzymes has been observed in many cancer cell lines, inhibition of these proteins may be of some interest in designing therapies for cancer treatment. There are four isoforms of UCH-L5 that exist as a result of alternative splicing events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UCH37
Applications Western Blot
Classification Monoclonal
Clone 854CT5.5.6
Concentration 0.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene UCHL5
Gene Accession No. Q9Y5K5
Gene Alias 5830413B11Rik; AD-019; CGI-70; INO80 complex subunit R; INO80R; ubiquitin carboxyl-terminal esterase L5; ubiquitin carboxyl-terminal hydrolase isozyme L5; ubiquitin carboxyl-terminal hydrolase L5; ubiquitin C-terminal hydrolase 37; ubiquitin C-terminal hydrolase L5; ubiquitin C-terminal hydrolase UCH37; ubiquitin thioesterase L5; UCH37; Uchl5; UCH-L5
Gene Symbols UCHL5
Host Species Mouse
Immunogen Purified His-tagged UCH37 protein.
Purification Method Protein G
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51377
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.