Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UCH37 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567383
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567383 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567383 Supplier Invitrogen™ Supplier No. PA567383
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MTGNAGEWCLM ESDPGVFTEL IKGFGCRGAQ VEEIWSLEPE NFEKLKPVHG LIFLFKWQPG EEPAGSVVQD SRLDT Highest antigen sequence identity to the following orthologs - mouse 95%, rat 96%.

UCH-L5 (ubiquitin carboxyl-terminal hydrolase isozyme L5), also known as UCH37, is a 329 amino acid protein that functions to edit polyubiquinated protein substrates. Since UCH-L5 has the potential to rescue ubiquinated proteins, including oncogenic proteins, from proteasomal degradation, it is likely that deregulation of UCH-L5 may affect tumor growth. Through associations with Smad7, UHC-L5 can dramatically upregulate TGFβ-dependent gene expression by de-ubiquinating and stabilizing TGFβ RI. Also, since overexpression of UCH-L5 and other deubiquitinating enzymes has been observed in many cancer cell lines, inhibition of these proteins may be of some interest in designing therapies for cancer treatment. There are four isoforms of UCH-L5 that exist as a result of alternative splicing events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UCH37
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UCHL5
Gene Accession No. Q9Y5K5
Gene Alias 5830413B11Rik; AD-019; CGI-70; INO80 complex subunit R; INO80R; ubiquitin carboxyl-terminal esterase L5; ubiquitin carboxyl-terminal hydrolase isozyme L5; ubiquitin carboxyl-terminal hydrolase L5; ubiquitin C-terminal hydrolase 37; ubiquitin C-terminal hydrolase L5; ubiquitin C-terminal hydrolase UCH37; ubiquitin thioesterase L5; UCH37; Uchl5; UCH-L5
Gene Symbols UCHL5
Host Species Rabbit
Immunogen Recombinant Human UCH37. Recombinant protein control fragment (Product #RP-105597).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51377
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.