Learn More
Invitrogen™ UCH37 Polyclonal Antibody

Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Specifications
Specifications
| Antigen | UCH37 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | UCHL5 |
| Gene Accession No. | Q9WUP7, Q9Y5K5 |
| Gene Alias | 5830413B11Rik; AD-019; CGI-70; INO80 complex subunit R; INO80R; ubiquitin carboxyl-terminal esterase L5; ubiquitin carboxyl-terminal hydrolase isozyme L5; ubiquitin carboxyl-terminal hydrolase L5; ubiquitin C-terminal hydrolase 37; ubiquitin C-terminal hydrolase L5; ubiquitin C-terminal hydrolase UCH37; ubiquitin thioesterase L5; UCH37; Uchl5; UCH-L5 |
| Gene Symbols | UCHL5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.