Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UCHL1+3 Rabbit anti-Human∣Mouse∣Rat, Polyclonal, Bioss
SDP

Catalog No. 501981575
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1575 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1575 Supplier Bioss Supplier No. BS11677R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The two ubiquitin C-terminal hydrolase (UCH) enzymes, UCHL1 and UCHL3, deubiquitinate ubiquitin-protein conjugates and control the cellular balance of ubiquitin. UCHL1 and UCHL3 are both small proteins of ˜220 amino acids that share more than 40% amino acid sequence identity. UCHL3 is universally expressed in all tissues, while UCHL1 is expressed exclusively in neuronal tissue, testis and ovary. The activity of UCHL3 is more than 200 fold higher than UCHL1 when a fluorogenic ubiquitin substrate is used. UCHL1 associates with monoubiquitin and UCHL3 binds to Nedd8, ubiquitin-like protein. UCHL1 and UCHL3 play a role in the regulation of neuronal development and spermatogenesis. UCHL1 is involved in the pathogenesis of Parkinson′s disease (PD) and Alzheimer′s disease (AD).

Specifications

Antigen UCHL1+3
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene UCHL1
Gene Accession No. P09936, Q00981, Q9R0P9
Gene Symbols UCHL1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human UCHL1+3 .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22223, 29545, 7345
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.