Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UCHL1 Mouse anti-Human, Monkey, Clone:7B8-G5-B8, Abnova™

Catalog No. 89333883 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-883 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-883 Supplier Abnova Corporation Supplier No. MAB17661

Mouse monoclonal antibody raised against recombinant human UCHL1.

The protein encoded by this gene belongs to the peptidase C12 family. This enzyme is a thiol protease that hydrolyzes a peptide bond at the C-terminal glycine of ubiquitin. This gene is specifically expressed in the neurons and in cells of the diffuse neuroendocrine system. Mutations in this gene may be associated with Parkinson disease

Specifications

Antigen UCHL1
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 7B8-G5-B8
Conjugate Unconjugated
Description ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase)
Dilution Immunocytochemistry (1:300)
Western Blot (1:1000)
The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.1M Tris-Glycine, 150 mM NaCl, pH 7.4 (50% glycerol, 0.2% sodium azide)
Gene UCHL1
Gene Alias PARK5, PGP9.5, Uch-L1
Host Species Mouse
Immunogen Recombinant protein corresponding to a fragment of human UCHL1 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 7345
Test Specificity This antibody detects endogenous levels of UCHL1/PGP9.5 and does not cross-react with related proteins.
Target Species Human, Monkey
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.