Learn More
Invitrogen™ UCHL3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533098
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Mouse, Rat, Dog, Elephant, Opossum, Turkey, Chicken, Lizard (100%); Orangutan, Monkey, Bovine, Horse, Rabbit, Pig, Salmon (94%); Pufferfish, Zebrafish, Stickleback, Water flea (89%); Xenopus (83%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
Regulatory associated protein of mTOR (RAPTOR) is a binding partner of target of rapamycin (TOR). It binds to 4EBP1 and p70alpha. The binding of raptor to mTOR is essential for the mTOR-catalyzed phosphorylation of 4EBP1 in vitro. It enhances the mTOR kinase activity toward p70alpha. It is also an essential scaffold for the mTOR-catalyzed phosphorylation of 4EBP1 and mediates TOR action in vivo.
Specifications
| UCHL3 | |
| Polyclonal | |
| Unconjugated | |
| UCHL3 | |
| RGD1561196; testicular tissue protein Li 221; ubiquitin carboxyl-terminal esterase L3 (ubiquitin thiolesterase); ubiquitin carboxyl-terminal hydrolase isozyme L3; ubiquitin C-terminal hydrolase L3; Ubiquitin thioesterase L3; ubiquitin thiolesterase; UCHL3; UCH-L3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 395626, 476947, 498560, 50933, 7347 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P15374, Q91Y78, Q9JKB1 | |
| UCHL3 | |
| Synthetic 18 amino acid peptide from internal region of human UCHL3. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat, Canine, Chicken | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.