Learn More
UCK1 Mouse anti-Human, Clone: OTI5E9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501543

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Phosphorylates uridine and cytidine to uridine monophosphate and cytidine monophosphate. Does not phosphorylate deoxyribonucleosides or purine ribonucleosides. Can use ATP or GTP as a phosphate donor. Can also phosphorylate cytidine and uridine nucleoside analogs such as 6-azauridine, 5-fluorouridine, 4- thiouridine, 5-bromouridine, N(4)-acetylcytidine, N(4)- benzoylcytidine, 5-fluorocytidine, 2-thiocytidine, 5- methylcytidine, and N(4)-anisoylcytidine.Specifications
UCK1 | |
Monoclonal | |
Unconjugated | |
UCK1 | |
URK1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
83549 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI5E9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9HA47 | |
UCK1 | |
Full length human recombit protein of human UCK1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.