Learn More
Invitrogen™ UCK1 Polyclonal Antibody

Description
Predicted to react with monkey based on sequence homology.
Specifications
Specifications
| Antigen | UCK1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | UCK1 |
| Gene Accession No. | P52623, Q9HA47 |
| Gene Alias | Cytidine monophosphokinase 1; UCK 1; Uck1; Umpk; uridine monophosphate kinase; uridine monophosphokinase 1; Uridine-cytidine kinase 1; URK1 |
| Gene Symbols | UCK1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.