Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UCKL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552101
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552101 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552101 Supplier Invitrogen™ Supplier No. PA552101
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PWYNEHGTQS KEAFAIGLGG GSASGKTTVA RMIIEALDVP WVVLLSMDSF YKVLTEQQQE QAAHNNFNFD HPDAFDFDLI ISTLKKLKQG KSVKVPIYDF TTHSRKKDWK TLYGANVI Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 96%.

Uridine kinase is a key enzyme in the pyrimidine salvage pathway of all mammalian cells. URKL1 (uridine kinase-like 1) is highly similar in structure and sequence to uridine kinase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UCKL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Uckl1
Gene Accession No. Q9NWZ5
Gene Alias 1110007H10Rik; F538; RGD1565465; UCK1L; UCK1-LIKE; Uckl1; uridine kinase-like 1; uridine-cytidine kinase 1 like 1; uridine-cytidine kinase 1-like 1; uridine-cytidine kinase-like 1; URKL1
Gene Symbols Uckl1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UCKL1. Recombinant protein control fragment (Product #RP-88610).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54963
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.