Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UCKL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118336
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118336 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5118336 Supplier Invitrogen™ Supplier No. PA5118336
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

UCKL1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP

Uridine kinase is a key enzyme in the pyrimidine salvage pathway of all mammalian cells. URKL1 (uridine kinase-like 1) is highly similar in structure and sequence to uridine kinase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UCKL1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene Uckl1
Gene Accession No. Q9NWZ5
Gene Alias 1110007H10Rik; F538; RGD1565465; UCK1L; UCK1-LIKE; Uckl1; uridine kinase-like 1; uridine-cytidine kinase 1 like 1; uridine-cytidine kinase 1-like 1; uridine-cytidine kinase-like 1; URKL1
Gene Symbols Uckl1
Host Species Rabbit
Immunogen E. coli-derived Human UCKL1 fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54963
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.