Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ UCLA GP130 2
GREENER_CHOICE

Catalog No. 658310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-831-0 10 mg
65-835-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 65-831-0 Supplier Tocris Bioscience™ Supplier No. 6583/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

gp130 agonist; brain penetrant and neuroprotectant

UCLA GP130 2 is a gp130 agonist. Protects against NMDA-induced toxicity in primary hippocampal neurons in vitro. Increases STAT3 phosphorylation in SH-SY5Y cells. Brain penetrant.

Specifications

Chemical Name or Material N-(4-Fluorophenyl)-4-phenyl-2-thiazolamine
CAS 339303-87-6
Quantity 10 mg
Molecular Formula C15H11FN2S
Formula Weight 270.32
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.