Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UCMA Antibody, Novus Biologicals™
SDP

Catalog No. NBP233469 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP233469 0.1 mL
NB436874 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP233469 Supplier Novus Biologicals Supplier No. NBP233469
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UCMA Polyclonal specifically detects UCMA in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen UCMA
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q8WVF2
Gene Alias C10orf49, chromosome 10 open reading frame 49, Gla-rich protein, GRP, unique cartilage matrix-associated protein, upper zone of growth plate and cartilage matrix associated
Gene Symbols UCMA
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: YYEEQRNEFENFVEEQNDEQEERSREAVEQWRQWHYDGLHPSYLYNRHHT
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221044
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.