Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UCMA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561295
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561295 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561295 Supplier Invitrogen™ Supplier No. PA561295
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YYEEQRNEFE NFVEEQNDEQ EERSREAVEQ WRQWHYDGLH PSYLYNRHHT Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.

UCMA is a gene that encodes for a protein that is specific to chondrocytes and highly charged. This protein is expressed abundantly in the upper immature zone of fetal and juvenile epiphyseal cartilage. The encoded protein undergoes proteolytic processing to generate a mature protein that is secreted into the extracellular matrix. The glutamic acid residues in the protein undergo gamma carboxylation in a vitamin K-dependent manner. Undercarboxylation of the protein has been associated with osteoarthritis in humans. Alternative splicing of the gene results in multiple transcript variants that encode different isoforms. UCMA is also associated with diseases such as Osteoarthritis and Poland Syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UCMA
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UCMA
Gene Accession No. Q8WVF2
Gene Alias 1110017I16Rik; AW121955; C10orf49; Gla-rich protein; GRP; GRP/UCMA; RGD1308977; Ucma; ucma {ECO:0000250; Ucma-C; UniProtKB:Q14BU0}; Unique cartilage matrix-associated protein; unique cartilage matrix-associated protein {ECO:0000250; Unique cartilage matrix-associated protein C-terminal fragment; upper zone of growth plate and cartilage matrix associated; upper zone of growth plate and cartilage matrix associated protein
Gene Symbols UCMA
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UCMA. Recombinant protein control fragment (Product #RP-100763).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221044
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.