Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UCN2 Rabbit anti-Human, Polyclonal, FabGennix

Catalog No. 502209575
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-9575 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-9575 Supplier Fabgennix Inc Supplier No. UCN2201AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UCN2 Polyclonal antibody specifically detects UCN2 in Human samples. It is validated for Dot Blot, ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen UCN2
Applications Dot Blot, ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene UCN2
Gene Accession No. Q96RP3
Gene Alias prepro-urocortin 2; SRP; Stresscopin-related peptide; ucn II; UCN2; Ucn-2; Ucn3; UCNI; ucn-II; UR; urocortin 2; urocortin 3; urocortin II; Urocortin-2; urocortin-related peptide; URP
Host Species Rabbit
Immunogen Synthetic peptide taken wtihin amino acid region 70-115 of human Urocortin 2 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90226
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.