Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UCN3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568437
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568437 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568437 Supplier Invitrogen™ Supplier No. PA568437
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UCN3 Polyclonal Antibody for Western Blot, IHC (P)

Urocortin 3 (UCN3) is a member of the corticotropin-releasing factor (CRF) family, encoded by the UCN3 gene, and is primarily involved in stress response and energy balance regulation. UCN3 is predominantly expressed in the hypothalamus and certain peripheral tissues, such as pancreatic islets, where it plays significant roles in modulating anxiety-related behaviors and metabolism. This neuropeptide functions by binding to CRF2 receptors, exerting effects on glucose metabolism and insulin secretion. UCN3 has been implicated in protecting the heart from ischemic damage and modulating the cardiovascular system's response to stress, highlighting its potential in treating cardiac disorders. Additionally, it influences nutrient partitioning and energy homeostasis, with expression levels affected by nutritional status. Research indicates that UCN3 deficiency may lead to impaired glucose tolerance and altered energy expenditure, demonstrating its importance in metabolic health. Furthermore, UCN3 has been associated with the regulation of neuroendocrine signaling pathways, providing insights into therapeutic strategies for metabolic and stress-related disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UCN3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene UCN3
Gene Accession No. Q969E3
Gene Alias prepro-urocortin 3; RGD1562213; SCP; SPC; Stresscopin; ucn III; Ucn3; UCNIII; urocortin 3; urocortin 3 (stresscopin); Urocortin II; Urocortin III; Urocortin-3
Gene Symbols UCN3
Host Species Rabbit
Immunogen Synthesized peptide derived from human UCN3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114131
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.