Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UCP1 Antibody (536435), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB6158P
Description
UCP1 Monoclonal antibody specifically detects UCP1 in Human, Mouse samples. It is validated for Western Blot, Western Blot, Flow Cytometry, ImmunocytochemistrySpecifications
UCP1 | |
Monoclonal | |
PE | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Flow Cytometry, Immunocytochemistry | |
536435 | |
Western Blot, Simple Western, Intracellular Staining by Flow Cytometry, Immunocytochemistry | |
SLC25A7mitochondrial brown fat uncoupling protein 1, Solute carrier family 25 member 7, thermogenin, UCP, UCP 1, uncoupling protein 1 (mitochondrial, proton carrier) | |
E. coli-derived recombinant human UCP1, Met1-Thr307, Accession # P25874 | |
0.1 mL | |
Cancer, Cellular Signaling, metabolism | |
7350 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction