Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UCP1 Monoclonal Antibody (GT1345)
GREENER_CHOICE

Catalog No. PIMA531535
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531535 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531535 Supplier Invitrogen™ Supplier No. MA531535
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (80%), Rat (80%), Rabbit (86%), Sheep (83%), Bovine (84%). Positive Control: mouse brown adipose. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Mitochondrial uncoupling proteins are members of the family of mitochondrial anion carrier proteins. UCPs separate oxidative phosphorylation from ATP synthesis with energy dissipated as heat, also referred to as the mitochondrial proton leak. UCPs facilitate the transfer of anions from the inner to the outer mitochondrial membrane and the return transfer of protons from the outer to the inner mitochondrial membrane. They also reduce the mitochondrial membrane potential in mammalian cells. Tissue specificity occurs for the different UCPs and the exact methods of how UCPs transfer H+/OH- are not known. UCPs contain the three homologous protein domains of MACPs. This gene is expressed only in brown adipose tissue, a specialized tissue which functions to produce heat.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UCP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone GT1345
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and no preservative
Gene Ucp1
Gene Accession No. P12242, P25874
Gene Alias AI385626; HGNC:12517; mitochondrial brown fat uncoupling protein 1; SLC25A7; solute carrier family 25 member 7; Thermogenin; Ucp; UCP 1; Ucp1; Ucpa; uncoupling protein; uncoupling protein 1; uncoupling protein 1 (mitochondrial, proton carrier); uncoupling protein 1, mitochondrial; uncoupling protein, mitochondrial; Uncp; unnamed protein product
Gene Symbols Ucp1
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the center region of human UCP1. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22227, 7350
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.