Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UCP2 Monoclonal Antibody (6C8B3)
GREENER_CHOICE

Catalog No. PIMA531945
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531945 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531945 Supplier Invitrogen™ Supplier No. MA531945
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31945 has been tested in indirect ELISA.

UCPs facilitate the transfer of anions from the inner to the outer mitochondrial membrane and the return transfer of protons from the outer to the inner mitochondrial membrane. They also reduce the mitochondrial membrane potential in mammalian cells. UCP2 gene is expressed in many tissues, with the greatest expression in skeletal muscle. UCP2 is thought to play a role in non shivering thermogenesis, obesity and diabetes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UCP2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 6C8B3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene Ucp2
Gene Accession No. P55851
Gene Alias BMIQ4; Mitochondrial uncoupling protein 2; Slc25a8; Solute carrier family 25 member 8; UCP 2; Ucp2; UCPH; uncoupling protein; uncoupling protein 2; uncoupling protein 2 (mitochondrial, proton carrier); uncoupling protein 2, mitochondrial
Gene Symbols Ucp2
Host Species Mouse
Immunogen Purified recombinant fragment of human UCP2 (AA: 1-309) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7351
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.