Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UCP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA124895
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA124895 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA124895 Supplier Invitrogen™ Supplier No. PA124895
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: human and mouse skeletal muscle, mitochondrial extract of rat brown adipose tissue. The corresponding sequence in rat and mouse UCP3 differ by 4 and 3 amino acids respectively. This antibody exhibits no cross reactivity with UCP1 or UCP2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The uncoupling protein UCP1 (formerly designated UCP) is an integral membrane protein unique to brown adipose tissue mitochondria. UCP1 forms a dimer that acts as a proton channel, which can uncouple oxidative phosphorylation by dissipating the electrochemical potential across the inner mitochondrial membrane. This process induces heat production in brown adipose tissue and is involved in regulation of body temperature and glucose metabolism. UCP2 is a structurally related protein that also uncouples mitochondrial respiration. It is more widely expressed in human and mouse tissues, including white adipose tissue and muscle, than is UCP1. UCP2 is thought to play a role in body weight regulation. An additional UCP family member, UCP3, is highly muscle specific and is possibly involved in the uncoupling of oxidative phosphorylation in skeletal muscle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UCP3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA and 15mM sodium azide; pH 7.4
Gene Ucp3
Gene Accession No. P55916, P56499, P56501
Gene Alias AI645527; mitochondrial uncoupling protein 3; Slc25a9; solute carrier family 25 member 9; UCP 3; UCP3; UCP-3; uncoupling protein; uncoupling protein 3; uncoupling protein 3 (mitochondrial, proton carrier); uncoupling protein 3, mitochondrial
Gene Symbols Ucp3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to human UCP-3 sequence (amino acids 254-267) with N-terminal lysine added, conjugated to KLH.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22229, 25708, 7352
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.