Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UCP5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP159598
Description
UCP5 Polyclonal specifically detects UCP5 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
UCP5 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
BMCP-1, BMCP1UCP 5, brain mitochondrial carrier protein 1, Mitochondrial uncoupling protein 5, solute carrier family 25 (mitochondrial carrier, brain), member 14, Solute carrier family 25 member 14, UCP5MGC149543 | |
Rabbit | |
21 kDa | |
100 μL | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
1 mg/ml | |
Western Blot 1.0 ug/ml, Immunocytochemistry/Immunofluorescence | |
Q5JY88 | |
SLC25A14 | |
Synthetic peptides corresponding to SLC25A14(solute carrier family 25 (mitochondrial carrier, brain), member 14) The peptide sequence was selected from the N terminal of SLC25A14 (CAI42439). Peptide sequence SIVAEFGTFPVDLTKTRLQVQGQSIDARFKEIKYRGMFHALFRICKEEGV The peptide sequence for this immunogen was taken from within the described region. | |
Protein A purified | |
RUO | |
9016 | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Sheep, Yeast, Zebrafish | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction