Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UCRC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542575
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542575 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542575 Supplier Invitrogen™ Supplier No. PA542575
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LFRRTSTFAL TIIVGVMFFE RAFDQGADAI YDHINEGKLW KHIKHKYENK Sequence homology: Cow: 93%; Dog: 86%; Guinea Pig: 86%; Human: 100%; Mouse: 93%; Rabbit: 86%; Rat: 100%.

UCRC is a subunit of mitochondrial complex III (ubiquinol-cytochrome c reductase; EC 1.10.2.2), which forms the middle segment of the respiratory chain of the inner mitochondrial membrane. UCRC is a subunit of mitochondrial complex III (ubiquinol-cytochrome c reductase; EC 1.10.2.2), which forms the middle segment of the respiratory chain of the inner mitochondrial membrane (Schagger et al., 1995 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen UCRC
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene UQCR10
Gene Accession No. Q9UDW1
Gene Alias 1110020P15Rik; AA960494; Complex III subunit 9; complex III subunit X; Cytochrome b-c1 complex subunit 9; cytochrome c1 non-heme 7 kDa protein; cytochrome C1, nonheme 7kDa protein; HSPC051; HSPC119; HSPC151; QCR9; ubiquinol-cytochrome c reductase complex (7.2 kD); ubiquinol-cytochrome c reductase complex 7.2 kDa protein; ubiquinol-cytochrome c reductase complex 7.2kDa protein; ubiquinol-cytochrome c reductase complex protein; ubiquinol-cytochrome c reductase, complex III subunit X; ubiquinol-cytochrome c reductase, complex III subunit X, 7.2kDa; UCCR7.2; Ucrc; Unknown (protein for MGC:127828); UQCR10
Gene Symbols UQCR10
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human UCRC.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29796
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.