Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UCRC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599025
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599025 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599025 Supplier Invitrogen™ Supplier No. PA599025
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UCRC Polyclonal Antibody for Western Blot, IHC (P), ELISA

UCRC is a subunit of mitochondrial complex III (ubiquinol-cytochrome c reductase; EC 1.10.2.2), which forms the middle segment of the respiratory chain of the inner mitochondrial membrane. UCRC is a subunit of mitochondrial complex III (ubiquinol-cytochrome c reductase; EC 1.10.2.2), which forms the middle segment of the respiratory chain of the inner mitochondrial membrane (Schagger et al., 1995 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen UCRC
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.44 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene UQCR10
Gene Accession No. Q9UDW1
Gene Alias 1110020P15Rik; AA960494; Complex III subunit 9; complex III subunit X; Cytochrome b-c1 complex subunit 9; cytochrome c1 non-heme 7 kDa protein; cytochrome C1, nonheme 7kDa protein; HSPC051; HSPC119; HSPC151; QCR9; ubiquinol-cytochrome c reductase complex (7.2 kD); ubiquinol-cytochrome c reductase complex 7.2 kDa protein; ubiquinol-cytochrome c reductase complex 7.2kDa protein; ubiquinol-cytochrome c reductase complex protein; ubiquinol-cytochrome c reductase, complex III subunit X; ubiquinol-cytochrome c reductase, complex III subunit X, 7.2kDa; UCCR7.2; Ucrc; Unknown (protein for MGC:127828); UQCR10
Gene Symbols UQCR10
Host Species Rabbit
Immunogen Recombinant Human Cytochrome b-c1 complex subunit 9 protein (1-63AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29796
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.