Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UD2A1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571376
Description
UD2A1 Polyclonal Antibody for Western Blot
The protein encoded by this gene belongs to the UDP-glycosyltransferase family, members of which catalyze biotransformation reactions in which lipophilic substrates are conjugated with glucuronic acid to increase water solubility and enhance excretion. They are of major importance in the conjugation and subsequent elimination of potentially toxic xenobiotics and endogenous compounds. This enzyme is expressed in the olfactory neuroepithelium, which lines the posterior nasal cavity and is exposed to a wide range of odorants and airborne toxic compounds. Hence, this protein has been suggested to be involved in clearing lipophilic odorant molecules from the sensory epithelium. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. This gene shares exon structure with the UDP glucuronosyltransferase 2A2 family member, which encodes N-terminally distinct isoforms.
Specifications
UD2A1 | |
Polyclonal | |
Unconjugated | |
UGT2A1 | |
UDP glucuronosyltransferase 2 family, polypeptide A1, complex locus; UDP glucuronosyltransferase family 2 member A1 complex locus; UDP glycosyltransferase 2 family, polypeptide A1; UDP-glucuronosyltransferase 2A1; UDPGT 2A1; UDPGT2A1; UGT2A1; UGT2A2; uridine diphosphate glycosyltransferase 2 family, member A1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
10941 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9Y4X1 | |
UGT2A1 | |
synthetic peptide directed towards the middle region of human UD2A1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction