Learn More
Description
Includes
Incorporate or detect GalNAc without expensive, specialized equipment!
Applications:
- For in vitro enzymatic incorporation of azido-sugars into specific, targeted glycans.
- Detect the presence or absence of GalNAC modifications.
Specifications
Specifications
| Quantity | 100 μg |
| Product Line | R&D Systems |
| Molecular Formula | C17H24N6O17P2 |
| Formula Weight | 646.35 Da |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
