Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ UDP disodium salt
GREENER_CHOICESDP

Catalog No. 311150 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-115-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 31-115-0 Supplier R&D Systems™ Supplier No. 3111/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Endogenous ligand; competitive antagonist at P2Y14 receptors

Endogenous P2Y receptor agonist which preferentially activates P2Y6. Shown to be a competitive antagonist at P2Y14 receptors.

Chemical Identifiers

CAS 27821-45-0
Molecular Formula C9H12N2Na2O12P2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 448.12
Purity >97%
Molecular Weight (g/mol) 448.12
Chemical Name or Material UDP disodium salt
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.