Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UDP glucose dehydrogenase Mouse anti-Human, Mouse, Clone: 2G11, Novus Biologicals™

Mouse Monoclonal Antibody
$340.50 - $499.50
Specifications
Antigen | UDP glucose dehydrogenase |
---|---|
Clone | 2G11 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000, Flow Cytometry 1:10-1:1062, ELISA, Immunocytochemistry/Immunofluorescence 1:100 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25946150
![]() |
Novus Biologicals
NBP25946150UL |
50 μL |
Each for $340.50
|
|
|||||
NBP259461X
![]() |
Novus Biologicals
NBP259461100UL |
100 μL |
Each for $499.50
|
|
|||||
Description
UDP glucose dehydrogenase Monoclonal specifically detects UDP glucose dehydrogenase in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
UDP glucose dehydrogenase | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Cancer, Core ESC Like Genes, Stem Cell Markers | |
Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
7358 | |
Recombinant human UGDH (1-494aa) purified from E. coli | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
2G11 | |
Western Blot 1:1000, Flow Cytometry 1:10-1:1062, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
EC 1.1.1.22, GDH, UDPGDHUDP-glucose dehydrogenase, UDP-Glc dehydrogenase, UDP-GlcDH, UDP-glucose 6-dehydrogenase, UGD, uridine diphospho-glucose dehydrogenase | |
UGDH | |
IgG2b κ | |
Protein A purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title