Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UEVLD Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305953R
Description
UEVLD Polyclonal specifically detects UEVLD in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
UEVLD | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Attp, EV and lactate/malate dehydrogenase domain-containing protein, signaling molecule ATTP, ubiquitin-conjugating enzyme E2 variant 3, UEV and lactate/malate dehyrogenase domains, UEV-3, UEV3FLJ11068 | |
Produced in rabbits immunized with E. coli-derived Human UEVLD fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
55293 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction