Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UEVLD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118337
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118337 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5118337 Supplier Invitrogen™ Supplier No. PA5118337
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

UEVLD Polyclonal Antibody for Western Blot, IHC (P), IP

Possible negative regulator of polyubiquitination.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UEVLD
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene UEVLD
Gene Accession No. Q8IX04
Gene Alias 8430408E05Rik; Attp; EV and lactate/malate dehydrogenase domain-containing protein; signaling molecule ATTP; ubiquitin-conjugating enzyme E2 variant 3; UEV and lactate/malate dehyrogenase domains; Uev3; UEV-3; UEVLD
Gene Symbols UEVLD
Host Species Rabbit
Immunogen E. coli-derived Human UEVLD fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55293
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.