Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UFC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555962
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555962 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555962 Supplier Invitrogen™ Supplier No. PA555962
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MADEATRRVV SEIPVLKTNA GPRDRELWVQ RLKEEYQSLI RYVENNKNAD NDWFRLESNK EGTRWFGKCW YIHDLLKYEF DIE Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 99%.

UFC1 is an E2-like conjugating enzyme for ubiquitin-fold modifier-1 (Komatsu et al., 2004 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen UFC1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UFC1
Gene Accession No. Q6BBI8, Q9CR09, Q9Y3C8
Gene Alias 1110021H02Rik; AA062310; CGI-126; ESTM29; HSPC155; ubiquitin-fold modifier conjugating enzyme 1; ubiquitin-fold modifier-conjugating enzyme 1; UFC1; ufc1-s; Ufm1-conjugating enzyme 1; Ufm1-conjugating enzyme 1-
Gene Symbols UFC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UFC1. Recombinant protein control fragment (Product #RP-94272).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 445268, 51506, 66155
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.