Learn More
Invitrogen™ UFC1 Recombinant Rabbit Monoclonal Antibody (8V0X7)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA555589

Description
Immunogen Sequence: MADEATRRVV SEIPVLKTNA GPRDRELWVQ RLKEEYQSLI RYVENNKNAD NDWFRLESNK EGTRWFGKCW YIHDLLKYEF DIEFDIPITY PTTAPEIAVP ELDGKTAKMY RGGKICLTDH FKPLWARNVP.
UFC1 is an E2-like conjugating enzyme for ubiquitin-fold modifier-1 (Komatsu et al., 2004 ).
Specifications
UFC1 | |
Recombinant Monoclonal | |
2 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6BBI8, Q9CR09, Q9Y3C8 | |
UFC1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-130 of Human UFC1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot | |
8V0X7 | |
Unconjugated | |
UFC1 | |
1110021H02Rik; AA062310; CGI-126; ESTM29; HSPC155; ubiquitin-fold modifier conjugating enzyme 1; ubiquitin-fold modifier-conjugating enzyme 1; UFC1; ufc1-s; Ufm1-conjugating enzyme 1; Ufm1-conjugating enzyme 1- | |
Rabbit | |
Protein A | |
RUO | |
445268, 51506, 66155 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.