Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UFC1 Recombinant Rabbit Monoclonal Antibody (JE65-12)
GREENER_CHOICE

Catalog No. PIMA544697
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544697 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544697 Supplier Invitrogen™ Supplier No. MA544697
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

UFC1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

UFC1 is an E2-like conjugating enzyme for ubiquitin-fold modifier-1 (Komatsu et al., 2004 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen UFC1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JE65-12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene UFC1
Gene Accession No. Q9Y3C8
Gene Alias 1110021H02Rik; AA062310; CGI-126; ESTM29; HSPC155; ubiquitin-fold modifier conjugating enzyme 1; ubiquitin-fold modifier-conjugating enzyme 1; UFC1; ufc1-s; Ufm1-conjugating enzyme 1; Ufm1-conjugating enzyme 1-
Gene Symbols UFC1
Host Species Rabbit
Immunogen Recombinant protein within human UFC1 aa 1-150/167.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51506
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.