Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UFD1L Antibody (OTI8G6), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274781AF350
Description
UFD1L Monoclonal specifically detects UFD1L in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
UFD1L | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
UFD1L | |
Full length recombinant protein of human UFD1L (NP_005650) produced in E.coli. | |
0.1 mL | |
7353.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI8G6 | |
Western Blot, Immunohistochemistry | |
ATP1C, ATP1G1ATPase, Na+/K+ transporting, gamma 1 polypeptide, FXYD domain containing ion transport regulator 2, FXYD domain-containing ion transport regulator 2, HOMG2, hypomagnesemia 2, renal, MGC12372, Na(+)/K(+) ATPase subunit gamma, Sodium pump gamma chain, sodium/potassium-transporting ATPase subunit gamma, Sodium-potassium-ATPase, gamma polypeptide | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction