Learn More
UFD1L Mouse anti-Human, Clone: OTI5E3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF804318

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene forms a complex with two other proteins, NPL4 and VCP, that is necessary for the degradation of ubiquitinated proteins. In addition, this complex controls the disassembly of the mitotic spindle and the formation of a closed nuclear envelope after mitosis. Mutations in this gene have been associated with Catch 22 syndrome as well as cardiac and craniofacial defects. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
UFD1L | |
Monoclonal | |
Unconjugated | |
UFD1 | |
UFD1L | |
Mouse | |
Affinity Chromatography | |
RUO | |
7353 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI5E3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q92890 | |
UFD1 | |
Full length human recombit protein of human UFD1L produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.