Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UFL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556501
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556501 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556501 Supplier Invitrogen™ Supplier No. PA556501
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QAALTKHLLK SVCTDITNLI FNFLASDLMM AVDDPAAITS EIRKKILSKL SEETKVALTK LHNSLNEKSI EDFISCLDSA AEACDIMVKR Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 88%.

UFL1 (UFM1 Specific Ligase 1) is a Protein Coding gene. Among its related pathways are Class I MHC mediated antigen processing and presentation and Innate Immune System. Gene Ontology (GO) annotations related to this gene include ligase activity and UFM1 transferase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UFL1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ufl1
Gene Accession No. O94874
Gene Alias 1810074P20Rik; AI429228; E3 UFM1-protein ligase 1; E3 UFM1-protein transferase 1; Kiaa0776; LZAP-binding protein; Maxer; mKIAA0776; Multiple alpha-helix protein located at ER; NLBP; Novel LZAP-binding protein; RCAD; Regulator of C53/LZAP and DDRGK1; Regulator of CDK5RAP3 and DDRGK1; RP3-393D12.1; UFL1; UFM1 specific ligase 1; UFM1-specific ligase 1
Gene Symbols Ufl1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UFL1. Recombinant protein control fragment (Product #RP-94597).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23376
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.