Learn More
UFL1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303456AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine
UFL1 (UFM1 Specific Ligase 1) is a Protein Coding gene. Among its related pathways are Class I MHC mediated antigen processing and presentation and Innate Immune System. Gene Ontology (GO) annotations related to this gene include ligase activity and UFM1 transferase activity.Specifications
UFL1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O94874, Q8CCJ3 | |
UFL1 | |
Between 744 and 794 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
UFL1 | |
KIAA0776, Maxer, NLBP, RCAD, RP3-393D12.1, E3 UFM1-protein ligase 1, LZAP-binding protein, Regulator of CDK5RAP3 and DDRGK1, novel LZAP-binding protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23376, 67490 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.