Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UFM1 Activating Enzyme/UBA5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | UFM1 Activating Enzyme/UBA5 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
UFM1 Activating Enzyme/UBA5 Polyclonal specifically detects UFM1 Activating Enzyme/UBA5 in Human samples. It is validated for Western Blot.Specifications
UFM1 Activating Enzyme/UBA5 | |
Polyclonal | |
Rabbit | |
Q9GZZ9 | |
79876 | |
Synthetic peptides corresponding to UBA5(ubiquitin-like modifier activating enzyme 5) The peptide sequence was selected from the middle region of UBA5. Peptide sequence VLSCVDNFEARMTINTACNELGQTWMESGVSENAVSGHIQLIIPGESACF. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
FLJ17281, FLJ23251UBA5, ThiFP1, UBE1DC1ubiquitin-activating enzyme E1 homolog, ubiquitin-like modifier activating enzyme 5, ubiquitin-like modifier-activating enzyme 5, UFM1-activating enzyme | |
UBA5 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title