Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UFM1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen UFM1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP179760
SDP
View Documents
Novus Biologicals
NBP179760
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

UFM1 Polyclonal specifically detects UFM1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

UFM1
Polyclonal
Rabbit
NP_057701
51569
Synthetic peptide directed towards the middle region of human UFM1The immunogen for this antibody is UFM1. Peptide sequence LSVPESTPFTAVLKFAAEEFKVPAATSAIITNDGIGINPAQTAGNVFLKH.
Primary
Western Blot
Unconjugated
RUO
bA131P10.1, BM-002, C13orf20, chromosome 13 open reading frame 20, ubiquitin-fold modifier 1
UFM1
IgG
9 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.