Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UFM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590754
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590754 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590754 Supplier Invitrogen™ Supplier No. PA590754
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse pancreas; Cellular Location: Cytoplasm, Nucleus.

UFM1 is a ubiquitin-like protein that is conjugated to target proteins by E1-like activating enzyme UBA5 (UBE1DC1; MIM 610552) and E2-like conjugating enzyme UFC1 (MIM 610554) in a manner analogous to ubiquitylation (see UBE2M; MIM 603173) (Komatsu et al, 2004).
TRUSTED_SUSTAINABILITY

Specifications

Antigen UFM1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene Ufm1
Gene Accession No. P61960, P61961, Q5BJP3
Gene Alias 1810045K17Rik; AI132708; AI463323; bA131P10.1; BM-002; C13orf20; ENSMUSG00000074598; Gm10726; hypothetical protein BM-002; RGD1304890; similar to ubiquitin-fold modifier 1; ubfm1; ubfm1 protein; ubiquitin fold modifier 1; ubiquitin fold modifier 1; ubiquitin-fold modifier 1; ubiquitin-fold modifier 1; ufm1; wu:fa10f05; wu:fb72g01; zBM-002; zgc:55335
Gene Symbols Ufm1
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 365797, 51569, 67890
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.