Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UFSP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555505
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555505 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555505 Supplier Invitrogen™ Supplier No. PA555505
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GSRDWIGCVE ASLCLAHFGG PQGRLCHVPR GVGLHGELER LYSHFAGGGG PVMVGGDADA RS Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 84%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen UFSP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UFSP1
Gene Accession No. Q6NVU6
Gene Alias inactive Ufm1-specific protease 1; UFM1 specific peptidase 1 (inactive); UFM1-specific peptidase 1 (inactive); UFM1-specific peptidase 1 (non-functional); UFSP; UfSP1
Gene Symbols UFSP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UFSP1. Recombinant protein control fragment (Product #RP-92802).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 402682
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.