Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UFSP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599002
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599002 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599002 Supplier Invitrogen™ Supplier No. PA599002
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UFSP2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Ubiquitin-fold modifier 1 (Ufm1)-specific protease 2 (UfSP2) is a cysteine protease that is responsible for the release of Ufm1 from Ufm1-conjugated cellular proteins, as well as for the generation of mature Ufm1 from its precursor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UFSP2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Ufsp2
Gene Accession No. Q99K23, Q9NUQ7
Gene Alias 1810047C23Rik; BHD; C4orf20; RGD1311161; UFM1 specific peptidase 2; UFM1-specific peptidase 2; ufm1-specific protease 2; UfSP2
Gene Symbols Ufsp2
Host Species Rabbit
Immunogen Recombinant Human Ufm1-specific protease 2 protein (63-343AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192169, 55325
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.