Learn More
Invitrogen™ UGCG Polyclonal Antibody

Description
Peptide sequence: LHLNKKATDK QPYSKLPGVS LLKPLKGVDP NLINNLETFF ELDYPKYEVL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 77%; Zebrafish: 100%.
Specifications
Specifications
| Antigen | UGCG |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | UGCG |
| Gene Accession No. | Q16739 |
| Gene Alias | AU043821; C80537; cb539; Ceramide glucosyltransferase; ectoplacental cone, invasive trophoblast giant cells, extraembryonic ectoderm and chorion sequence 21; Epcs21; GCS; GlcT; GLCT1; GLCT-1; glucosylceramide synthase; hypothetical protein LOC553944; sb:cb539; UDP-glucose ceramide glucosyltransferase; UDP-glucose:ceramide glycosyltransferase; UDP-glucose:N-acylsphingosine D-glucosyltransferase; Ugcg; Ugcgl; zgc:112506 |
| Gene Symbols | UGCG |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.