Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UGCG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554986
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554986 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554986 Supplier Invitrogen™ Supplier No. PA554986
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLHLNKKATD KQPYSKLPGV SLLKPLKGVD PNLINNLETF FELDYPKYEV LLCVQDHDDP AIDVCKKLLG KYPNVDARLF IGGKKVGINP KINNLMPGYE VAKYDLI Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 98%.

May serve as a flippase as well as a glucosyltransferase that transfers glucose to ceramide. It Catalyzes; UDP-glucose + Nacylsphingosine = UDP + D-glucosyl-N-acylsphingosine. Ceramide Glycosyltransferase is the first step in the Glycosphingolipid synthesis; first glycosylation step. Glucosylceramide synthase (GlcT) and lactosylceramide synthase (GalT) are key enzymes for the synthesis of major glycosphingolipids of vertebrates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UGCG
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UGCG
Gene Accession No. Q16739
Gene Alias AU043821; C80537; cb539; Ceramide glucosyltransferase; ectoplacental cone, invasive trophoblast giant cells, extraembryonic ectoderm and chorion sequence 21; Epcs21; GCS; GlcT; GLCT1; GLCT-1; glucosylceramide synthase; hypothetical protein LOC553944; sb:cb539; UDP-glucose ceramide glucosyltransferase; UDP-glucose:ceramide glycosyltransferase; UDP-glucose:N-acylsphingosine D-glucosyltransferase; Ugcg; Ugcgl; zgc:112506
Gene Symbols UGCG
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UGCG. Recombinant protein control fragment (Product #RP-93000).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7357
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.