Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UGDH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588347
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588347 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588347 Supplier Invitrogen™ Supplier No. PA588347
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EGTAIKDLKN PDRVLIGGDE TPEGQRAVQA LCAVYEHWVP REKILTTNTW SSELSKLAAN AFLAQRISSI NSISALCEAT GADVEEVATA IGMDQRIGNK FLKASVGFGG SCFQKDVLNL VYLCEALNLP EVARYWQQVI DMNDYQRRRF ASRIIDSLFN TVTDKKIAIL GFAFKKDTGD TRESSSIYIS KYLMDEGAHL HIYDPKVPRE QIVVDLSHPG VSEDDQVSRL VTISKDPYEA CDGAHAVVIC TEWDMFKELD YERIHKKMLK PAFIFDGRRV LDGLHNELQT IGFQIETIGK KVSSKRIPYA PSGEIPKFSL QDPPNKKPKV; Positive Samples: HeLa, THP-1, 22Rv1, ES-2, U-937, Mouse kidney, Mouse intestine.

The protein encoded by this gene converts UDP-glucose to UDP-glucuronate and thereby participates in the biosynthesis of glycosaminoglycans such as hyaluronan, chondroitin sulfate, and heparan sulfate. These glycosylated compounds are common components of the extracellular matrix and likely play roles in signal transduction, cell migration, and cancer growth and metastasis. The expression of this gene is up-regulated by transforming growth factor beta and down-regulated by hypoxia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UGDH
Applications ELISA, Western Blot, Western Blot
Classification Polyclonal
Concentration 0.44 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene UGDH
Gene Accession No. O60701, O70475
Gene Alias GDH; UDP UGD; UDPGDH; UDP-Glc dehydrogenase; UDP-GlcDH; UDP-glucose 6-dehydrogenase; UDP-glucose dehydrogeanse; UDP-glucose dehydrogenase; UGD; UGDH; uridine diphospho-glucose dehydrogenase
Gene Symbols UGDH
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 165-494 of human UGDH (NP_0033501).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22235, 7358
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.