Learn More
Invitrogen™ UGDH Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528170
Description
Recommended positive controls: HepG2, mouse liver, rat liver, UGDH-transfected 293T. Predicted reactivity: Mouse (97%), Rat (98%), Zebrafish (91%), Xenopus laevis (94%), Chicken (95%), Rhesus Monkey (98%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene converts UDP-glucose to UDP-glucuronate and thereby participates in the biosynthesis of glycosaminoglycans such as hyaluronan, chondroitin sulfate, and heparan sulfate. These glycosylated compounds are common components of the extracellular matrix and likely play roles in signal transduction, cell migration, and cancer growth and metastasis. The expression of this gene is up-regulated by transforming growth factor beta and down-regulated by hypoxia.
Specifications
| UGDH | |
| Polyclonal | |
| Unconjugated | |
| UGDH | |
| GDH; UDP UGD; UDPGDH; UDP-Glc dehydrogenase; UDP-GlcDH; UDP-glucose 6-dehydrogenase; UDP-glucose dehydrogeanse; UDP-glucose dehydrogenase; UGD; UGDH; uridine diphospho-glucose dehydrogenase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22235, 7358, 83472 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1.36 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| O60701, O70199, O70475 | |
| UGDH | |
| Recombinant protein encompassing a sequence within the center region of human UGDH. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.