Learn More
Invitrogen™ UGGT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA592309
Description
Immunogen sequence: PNNMIHQVPI KSLPQEWLWC ETWCDDASKK RAKTIDLCNN PMTKEPKLEA AVRIVPEWQD YDQEIKQLQI RFQKEKETGA LYKEKTKEPS REGPQKREEL; Positive Samples: SH-SY5Y, 293T, HepG2, HL-60, HT-1080, Mouse liver, Mouse kidney, Rat liver, Rat brain; Cellular Location: Endoplasmic reticulum lumen, Endoplasmic reticulum-Golgi intermediate compartment.
UDP-glucose:glycoprotein glucosyltransferase (UGGT) is a 170 kDa lumenal endoplasmic reticulum (ER) resident protein that catalyzes monoglucosylation of high mannose oligosaccharides. UGGT catalyzes the transfer of glucose from UDP-glucose to Man7-9GlcNAc2-moiety of unfolded glycoproteins in a Ca2+ dependent reaction. UGGT binds unfolded glycoprotein via two elements on its substrate: a N-acetylglucosamine unit on the oligosaccharide and hydrophobic amino acids that are exposed in the denatured conformation. It is postulated that UGGT serves as a sensor that is able to distinguish between folded and denatured protein in the ER.
Specifications
UGGT1 | |
Polyclonal | |
Unconjugated | |
Uggt1 | |
0910001L17Rik; A930007H10Rik; AA589501; AI414429; AI448372; C820010P03Rik; Gt; HUGT1; RUGT; rUGT1; UDP--Glc:glycoprotein glucosyltransferase; UDP-glucose ceramide glucosyltransferase-like 1; UDP-glucose glycoprotein glucosyltransferase 1; UDP-glucose glycoprotein: glucosyltransferase UGGT; UDP-glucose:glycoprotein glucosyltransferase; UDP-glucose:glycoprotein glucosyltransferase 1; UGCGL1; Uggt; Uggt1; Ugt1; Ugtr | |
Rabbit | |
Affinity Chromatography | |
RUO | |
171129, 320011, 56886 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.96 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6P5E4, Q9JLA3, Q9NYU2 | |
Uggt1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1456-1555 of human UGGT1 (NP_0645051). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.