Learn More
Invitrogen™ UGGT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113879
Description
Immunogen sequence: LCNNPKTKES KLKAAARIVP EWVEYDAEIR QLLDHLENKK QDTILTHDEL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Horse: 91%; Human: 100%; Mouse: 91%; Pig: 91%; Rabbit: 82%; Rat: 100%; Yeast: 90%.
UDP-glucose:glycoprotein glucosyltransferase (UGT) is a soluble protein of the endoplasmic reticulum (ER) that selectively reglucosylates unfolded glycoproteins, thus providing quality control for protein transport out of the ER.
Specifications
| UGGT2 | |
| Polyclonal | |
| Unconjugated | |
| UGGT2 | |
| HUGT2; UDP-Glc:glycoprotein glucosyltransferase 2; UDP--Glc:glycoprotein glucosyltransferase 2; UDP-glucose ceramide glucosyltransferase-like 1; UDP-glucose ceramide glucosyltransferase-like 2; UDP-glucose glycoprotein glucosyltransferase 2; UDP-glucose:glycoprotein glucosyltransferase 2; UGCGL2; UGGT2; UGT2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 55757 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NYU1 | |
| UGGT2 | |
| Synthetic peptide directed towards the middle region of human UGCGL2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.