Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UGGT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA527960
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527960 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527960 Supplier Invitrogen™ Supplier No. PA527960
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

UDP-glucose:glycoprotein glucosyltransferase (UGT) is a soluble protein of the endoplasmic reticulum (ER) that selectively reglucosylates unfolded glycoproteins, thus providing quality control for protein transport out of the ER.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UGGT2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene UGGT2
Gene Accession No. Q9NYU1
Gene Alias HUGT2; UDP-Glc:glycoprotein glucosyltransferase 2; UDP--Glc:glycoprotein glucosyltransferase 2; UDP-glucose ceramide glucosyltransferase-like 1; UDP-glucose ceramide glucosyltransferase-like 2; UDP-glucose glycoprotein glucosyltransferase 2; UDP-glucose:glycoprotein glucosyltransferase 2; UGCGL2; UGGT2; UGT2
Gene Symbols UGGT2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human UGCGL2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55757
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.