Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UGI Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144524
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144524 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144524 Supplier Invitrogen™ Supplier No. PA5144524
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UGI Polyclonal Antibody for Western Blot, ELISA

This protein binds specifically and reversibly to the host uracil-DNA glycosylase, preventing removal of uracil residues from PBS2 DNA by the host uracil-excision repair system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UGI
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene Alias Uracil-DNA glycosylase inhibitor UGI
Host Species Rabbit
Immunogen Recombinant Bacillus phage PBS2 (Bacteriophage PBS2) UGI protein.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.