Learn More
Stoelting™ Ugo Basile Treadmill
SDP
Stoelting™ Ugo Basile Treadmill is used to study the impact of physical activity on health
$29250.00
Specifications
For Use With (Application) Neuroscience
Animal Rodent
View More Specs
Products 2
Catalog Number Mfr. No. Model Price Quantity & Availability  
Catalog Number Mfr. No. Model Price Quantity & Availability  
10-000-998
SDP
View Documents
Stoelting™
57630
Treadmill
Each for $29,250.00
Only null left
Add to Cart
10-000-999
SDP
View Documents
Stoelting™
57633
Treadmill, tethered rodent
Each for $29,250.00
Only null left
Add to Cart
Description
Description
  • Adjustable speed and slope
  • Contant, accelerating or custom modes
  • Shock grid is incorporated at the end of the lanes should an aversive stimulus be required.
Specifications
Specifications
Neuroscience
Rodent
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.