Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UGRP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120195
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120195 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5120195 Supplier Invitrogen™ Supplier No. PA5120195
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive test controls include: A-549, Raji, mouse lung, rat lung. The target is usually found in the following locations: Secreted. Immunogen sequence: MKLVTIFLLV TISLCSYSAT AFLINKVPLP VDKLAPLPLD NILPFMDPLK LLLKTLGISV EHLVEGLRKC VNELGPEASE AVKKLLEALS HLV.

TRUSTED_SUSTAINABILITY

Specifications

Antigen UGRP1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.66 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene SCGB3A2
Gene Accession No. Q920H1, Q96PL1
Gene Alias LU103; Pneumo secretory protein 1; PNSP1; PnSP-1; SCGB3A2; Secretoglobin family 3A member 2; secretoglobin, family 3A, member 2; UGRP1; UGRP1pnSP-1; UNQ566/PRO1128; Uteroglobin-related protein 1
Gene Symbols SCGB3A2
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117156, 117158, 498854
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.